missing translation for 'onlineSavingsMsg'
Learn More

trans-Trimethoxyresveratrol, MedChemExpress

Product Code. 16424917
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16424917 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16424917 Supplier Medchem Express Supplier No. HYN1408
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Trans-Trimethoxyresveratrol is a derivative of Resveratrol (RSV),and it may be a more potent anti-inflammatory, antiangiogenic and vascular-disrupting agent when compared with resveratrol.

Trans-Trimethoxyresveratrol is a derivative of Resveratrol (RSV),and it may be a more potent anti-inflammatory, antiangiogenic and vascular-disrupting agent when compared with resveratrol.

Specifications

Chemical Name or Material trans-Trimethoxyresveratrol
CAS 22255-22-7
Color White
Purity Grade Notes Research
Molecular Formula C17H18O3
Quantity 50 mg
Synonym trans-trismethoxy Resveratrol E-Resveratrol Trimethyl Ether Tri-O-methylresveratrol
Solubility Information DMSO : ≥ 50 mg/mL (184.97 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES COC1=CC=C(/C=C/C2=CC(OC)=CC(OC)=C2)C=C1
Molecular Weight (g/mol) 270.32
Formula Weight 270.32
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.