missing translation for 'onlineSavingsMsg'
Learn More

trans,trans,trans-1,5,9-Cyclododecatrene, TRC

Product Code. 30425739
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30425739 5 g 5g
30424233 500 mg -
30422440 1 g -
3 options
This item is not returnable. View return policy

Product Code. 30425739

Brand: Toronto Research Chemicals TRCT2211605G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 676-22-2
Formula Weight 162.1409
InChI Formula InChI=1S/C12H18/c1-2-4-6-8-10-12-11-9-7-5-3-1/h1-2,7-10H,3-6,11-12H2/b2-1+,9-7+,10-8+
IUPAC Name (1E,5E,9E)-cyclododeca-1,5,9-triene
Molecular Formula C12H18
Molecular Weight (g/mol) 162.27
SMILES C1C\C=C\CC\C=C\CC\C=C\1
Synonym (1E,5E,9E)-1,5,9-Cyclododecatriene, (1E,5E,9E)-Cyclododeca-1,5,9-triene, (E,E,E)-1,5,9-Cyclododecatriene, all-trans-1,5,9-Cyclododecatriene
Chemical Name or Material trans,trans,trans-1,5,9-Cyclododecatrene

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.