missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-Muconic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | trans,trans-Muconic Acid |
| CAS | 3588-17-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6 H6 O4 |
| Quantity | 10 g |
| Synonym | 2,4-Hexadienedioic acid','(2E,4E)- (9CI','ACI), (2E,4E)-2,4-Hexadienedioic acid (ACI), 2,4-Hexadienedioic acid','(E,E)- (8CI), (E,E)-1,3-Butadiene-1,4-dicarboxylic acid, (E,E)-Muconic acid, NSC 66486, trans,trans-2,4-Hexadienedioic acid, trans,trans-Muconic acid |
| InChI Formula | InChI=1S/C6H6O4/c7-5(8)3-1-2-4-6(9)10/h1-4H,(H,7,8)(H,9,10)/b3-1+,4-2+ |
| SMILES | OC(=O)\C=C\C=C\C(=O)O |
| IUPAC Name | (2E,4E)-hexa-2,4-dienedioic acid |
| Molecular Weight (g/mol) | 142.11 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?