missing translation for 'onlineSavingsMsg'
Learn More

trans,trans Farnesyl Thiosalicylic Acid, TRC

Product Code. 30389121
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389121 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389121 Supplier Toronto Research Chemicals Supplier No. TRCF102500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans, trans Farnesylthiosalicylic Acid
CAS 162520-00-5
Molecular Formula C22 H30 O2 S
Quantity 100 mg
InChI Formula InChI=1S/C22H30O2S/c1-17(2)9-7-10-18(3)11-8-12-19(4)15-16-25-21-14-6-5-13-20(21)22(23)24/h5-6,9,11,13-15H,7-8,10,12,16H2,1-4H3,(H,23,24)/b18-11+,19-15+
SMILES CC(=CCC\C(=C\CC\C(=C\CSc1ccccc1C(=O)O)\C)\C)C
IUPAC Name 2-[(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl]sulfanylbenzoic acid
Molecular Weight (g/mol) 358.54
Formula Weight 358.1967
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.