Learn More
trans,trans-Farnesol, 97%
Description
trans,trans-Farnesol is an intermediate used in the biological synthesis of cholesterol from mevalonic acid. It is used in the perfume industry. Further, it is used in the preparation of terpenes such as squalenes, cembranoids and forskolin. It is also used in the synthesis of Cecropia juvenile hormone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
trans,trans-Farnesol is an intermediate used in the biological synthesis of cholesterol from mevalonic acid. It is used in the perfume industry. Further, it is used in the preparation of terpenes such as squalenes, cembranoids and forskolin. It is also used in the synthesis of Cecropia juvenile hormone.
Solubility
Miscible with alcohol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Density | 0.886 |
| Boiling Point | 137°C (3 mmHg) |
| Flash Point | 96°C (204°F) |
| Refractive Index | 1.49 |
| Quantity | 5 g |
| Beilstein | 1723039 |
| Sensitivity | Light sensitive |
| Merck Index | 14,3937 |
| Solubility Information | Miscible with alcohol. Immiscible with water. |
| Formula Weight | 222.37 |
| Show More |
Safety and Handling
- trans,trans-Farnesol
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : 3,7,11-trimethyl-2,6,10-Dodecatrien-1-ol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.