missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-2,4-Undecadienal, 90+%, remainder mainly trans,cis isomer

Product Code. 11445453
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11445453

Brand: Thermo Scientific Alfa Aesar A13419.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30361-29-6 | C11H18O | 166.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30361-29-6
Molecular Formula C11H18O
Molecular Weight (g/mol) 166.264
MDL Number MFCD00014677
InChI Key UVIUIIFPIWRILL-XBLVEGMJSA-N
Synonym 2,4-undecadienal, 2e,4e-undeca-2,4-dienal, undeca-2,4-dien-1-al, trans,trans-2,4-undecadien-1-al, 2,4-undecadienal, 2e,4e, 2,4-trans,trans-undecadienal, unii-7l26s7bw06, 2,4-undecadienal, e,e, trans,trans-2,4-undecadienal, e,e-2,4-undecadien-1-al
PubChem CID 5367531
IUPAC Name (2E,4E)-undeca-2,4-dienal
SMILES CCCCCCC=CC=CC=O

Specifications

Density 0.87
Boiling Point 128°C to 129°C (10 mmHg)
Flash Point 107°C (225°F)
Refractive Index 1.512
Quantity 5 g
Beilstein 1706330
Sensitivity Air sensitive
Formula Weight 166.27
Percent Purity ≥90%
Assay remainder mainly trans,cis isomer
Chemical Name or Material trans,trans-2,4-Undecadienal
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.