missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-2,4-Hexadienyl acetate, 97%, Thermo Scientific™

Product Code. 11357516
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357516 25 g 25g
11347516 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357516 Supplier Thermo Scientific Alfa Aesar Supplier No. L11050.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
trans,trans-2,4-Hexadienyl acetate is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1516-17-2
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
MDL Number MFCD00036521
InChI Key PXVKYPFROMBALG-VNKDHWASSA-N
Synonym sorbyl acetate, 2,4-hexadienyl acetate, trans,trans-2,4-hexadienyl acetate, hexa-2,4-dienyl acetate, 2,4-hexadien-1-ol, acetate, unii-5416sj1t8t, acetic acid, 2,4-hexadien-1-ol ester, acetic acid, 2,4-hexadienyl ester, 2,4-hexadien-1-ol, 1-acetate, 2,4-hexadienylester kyseliny octove
PubChem CID 5363491
IUPAC Name [(2E,4E)-hexa-2,4-dienyl] acetate
SMILES CC=CC=CCOC(=O)C

Specifications

Density 0.93
Boiling Point 100°C (20 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.4767
Quantity 25 g
Beilstein 1702607
Solubility Information Insoluble in water.
Formula Weight 140.18
Percent Purity 97%
Chemical Name or Material trans,trans-2,4-Hexadienyl acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.