missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-2,4-Hexadien-1-ol, 99%, stabilized

Product Code. 10063560 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10063560 50 g 50g
10701361 10 g 10g
11984621 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10063560 Supplier Thermo Scientific Acros Supplier No. 325570500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17102-64-6 | C6H10O | 98.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17102-64-6
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00002925
InChI Key MEIRRNXMZYDVDW-MQQKCMAXSA-N
Synonym sorbic alcohol, 2,4-hexadien-1-ol, sorbyl alcohol, trans,trans-2,4-hexadien-1-ol, 2,4-hexadienol, hexacose, hexakose, sorbinic alcohol, 1-hydroxy-2,4-hexadiene, 2e,4e-hexa-2,4-dien-1-ol
PubChem CID 641256
IUPAC Name (2E,4E)-hexa-2,4-dien-1-ol
SMILES C\C=C\C=C\CO

Specifications

Melting Point 30.5°C to 31.5°C
Color Colorless to Yellow
Density 0.8700g/mL
Boiling Point 80.0°C (12.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=CHCH=CHCH2OH
Refractive Index 1.4990 to 1.5010
Quantity 50 g
Merck Index 15,8848
Specific Gravity 0.87
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and oils
Stabilizer 0.1% alpha-tocopherol
Formula Weight 98.14
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material trans, trans-2, 4-Hexadien-1-ol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • trans,trans-2,4-Hexadien-1-ol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.