missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-2,4-Decadienal, 90+%, remainder mainly trans,cis isomer, Thermo Scientific™

Product Code. p-7044380
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11352376

Brand: Thermo Scientific Alfa Aesar L06103.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25152-84-5
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00007007
InChI Key JZQKTMZYLHNFPL-BLHCBFLLSA-N
Synonym 2e,4e-deca-2,4-dienal, 2,4-decadienal, e,e-2,4-decadienal, trans,trans-2,4-decadienal, 2,4-decadienal, 2e,4e, 2e,4e-decadienal, 2e,4e-2,4-decadienal, 2,4-decadienal, e,e, 2-trans-4-trans-decadienal, ccris 4029
PubChem CID 5283349
IUPAC Name (2E,4E)-deca-2,4-dienal
SMILES CCCCCC=CC=CC=O

Specifications

Density 0.872
Boiling Point 110°C (7 mmHg)
Flash Point 101°C (213°F)
Odor Sweet
Refractive Index 1.516
Quantity 1 g
Beilstein 1704897
Sensitivity Air sensitive
Formula Weight 152.24
Percent Purity ≥90%
Assay remainder mainly trans, cis isomer
Chemical Name or Material trans,trans-2,4-Decadienal
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.