Learn More
trans,trans-2,4-Decadien-1-ol, 90%, remainder mainly trans, cis isomer
Description
trans,trans-2,4-Decadien-1-ol is used as food flavor and as an intermediate in organic chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
trans,trans-2,4-Decadien-1-ol is used as food flavor and as an intermediate in organic chemical synthesis.
Solubility
Not miscible in water. Soluble in fat.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 0.87 |
| Boiling Point | 80°C (0.5 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.489 |
| Quantity | 1 g |
| Solubility Information | Not miscible in water. Soluble in fat. |
| Formula Weight | 154.26 |
| Percent Purity | 90% |
| Assay | remainder mainly trans, cis isomer |
| Chemical Name or Material | trans,trans-2,4-Decadien-1-ol |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.