missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-1,4-Diphenyl-1,3-butadiene, 98+%

Product Code. 11438157
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438157 5 g 5g
11448157 25 g 25g
11458157 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438157 Supplier Thermo Scientific Alfa Aesar Supplier No. A17531.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 538-81-8 | C16H14 | 206.29 g/mol

trans,trans-1,4-Diphenyl-1,3-butadiene reacts with Rieke metal complexes of barium and strontium to prepare metal-diene reagents, which is used for the carbocyclization with dichloroalkanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans,trans-1,4-Diphenyl-1,3-butadiene reacts with Rieke metal complexes of barium and strontium to prepare metal-diene reagents, which is used for the carbocyclization with dichloroalkanes.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-81-8
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00004791
InChI Key JFLKFZNIIQFQBS-FNCQTZNRSA-N
Synonym bistyryl, 1,4-diphenyl-1,3-butadiene, 1,4-diphenylbutadiene, trans,trans-1,4-diphenyl-1,3-butadiene, 1,4-diphenylerythrene, distyryl, trans,trans-1,4-diphenylbuta-1,3-diene, 1,4-diphenylbuta-1,3-diene, 1,1'-1e,3e-buta-1,3-diene-1,4-diyldibenzene, 1,3-butadiene, 1,4-diphenyl
PubChem CID 641683
ChEBI CHEBI:35100
IUPAC Name [(1E,3E)-4-phenylbuta-1,3-dienyl]benzene
SMILES C(\C=C\C1=CC=CC=C1)=C/C1=CC=CC=C1

Specifications

Melting Point 151°C to 154°C
Boiling Point 350°C (720 mmHg)
Odor Odorless
Quantity 5 g
Beilstein 1905937
Merck Index 14,3320
Solubility Information Slightly soluble in water.
Formula Weight 206.29
Percent Purity ≥98%
Chemical Name or Material trans,trans-1,4-Diphenyl-1,3-butadiene
compliance-icons
Product Identifier
  • trans,trans-1,4-Diphenyl-1,3-butadiene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: trans,trans-1,4-Diphenyl-1,3-butadiene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.