missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-Stilbene-d10 (rings-d10), CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | trans-Stilbene-D10 (diphenyl-D10) |
| CAS | 20748-24-7 |
| Molecular Formula | C14 D10 H2 |
| Quantity | 0.5 g |
| Synonym | Benzene-1,2,3,4,5-d5, 6,6'-(1 E)-1,2-ethenediylbis- (ACI), 6,6'-(1 E)-1,2-Ethenediylbis[benzene-1,2,3,4,5-d5] (ACI), Benzene-d5, 6,6'-(1,2-ethenediyl)bis-, (E)- (ZCI), Benzene-d5, 6,6'-(1 E)-1,2-ethenediylbis- (9 CI), Stilbene-2,2',3,3',4,4',5,5',6,6'-d10, (E)- (8 CI), trans-Stilbene-2,2',3,3',4,4',5,5',6,6'-d10, trans-Stilbene-2,2',3,3',4,4',5,5',6,6'-d10, trans-Stilbene-2,2',3,3',4,4',5,5',6,6'-d10 |
| InChI Formula | InChI=1 S/C14H12/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-12 H/b12-11+/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(\C=C\c2c([2 H])c([2 H])c([2 H])c([2 H])c2[2 H])c([2 H])c1[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-[(E)-2-(2,3,4,5,6-pentadeuteriophenyl)ethenyl]benzene |
| Molecular Weight (g/mol) | 190.307 |
| Formula Weight | 190.157 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction