missing translation for 'onlineSavingsMsg'
Learn More

trans-Stilbene, 98%

Product Code. 11433933
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433933 2500 g 2500g
11413933 100 g 100g
11423933 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433933 Supplier Thermo Scientific Alfa Aesar Supplier No. A10232.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-30-0 | C14H12 | 180.25 g/mol

trans-Stilbene is used in the manufacturing of dye lasers, optical brighteners, non-steroidal synthetic estrogens such as diethylstilbestrol, fosfestrol and dienestrol. It is used as a phosphor and a scintillator. It is also involved in transition metal catalyzed asymmetric epoxidation and dihydroxylation. Further, it is used as raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Stilbene is used in the manufacturing of dye lasers, optical brighteners, non-steroidal synthetic estrogens such as diethylstilbestrol, fosfestrol and dienestrol. It is used as a phosphor and a scintillator. It is also involved in transition metal catalyzed asymmetric epoxidation and dihydroxylation. Further, it is used as raw material in organic synthesis.

Solubility
Soluble in benzene and ether. Insoluble in water.

Notes
Air and light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-30-0
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
MDL Number MFCD00064300
InChI Key PJANXHGTPQOBST-VAWYXSNFSA-N
Synonym trans-stilbene, e-stilbene, stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, bibenzylidene, bibenzylidine, bibenzal, trans-1,2-diphenylethene, alpha,beta-diphenylethylene
PubChem CID 638088
ChEBI CHEBI:36007
IUPAC Name (E)-stilbene
SMILES C1=CC=C(C=C1)C=CC2=CC=CC=C2

Specifications

Melting Point 122°C to 125°C
Boiling Point 305°C to 307°C
Odor Odorless
Quantity 2500 g
UN Number UN3077
Beilstein 1616740
Merck Index 14,8817
Formula Weight 180.25
Percent Purity 98%
Chemical Name or Material trans-Stilbene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.