missing translation for 'onlineSavingsMsg'
Learn More

trans-Piceatannol, TRC

Product Code. 30003371
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003371 10 mg 10mg
30011632 1 mg 1mg
30009513 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003371 Supplier Toronto Research Chemicals Supplier No. TRCP43630010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10083-24-6
Molecular Formula C14 H12 O4
Molecular Weight (g/mol) 244.24
Synonym Piceatannol, 1,2-Benzenediol, 4-[2-(3,5-dihydroxyphenyl)ethenyl]-, (E)-, 3,3',4,5'-Stilbenetetrol, (E)- (8CI), Piceatannol (6CI), 4-[(1E)-2-(3,5-Dihydroxyphenyl)ethenyl]-1,2-benzenediol, (E)-Piceatannol, Astringenin, trans-3,3',4,5'-Tetrahydroxystilbene, trans-Piceatannol
IUPAC Name 4-[(E)-2-(3,5-dihydroxyphenyl)ethenyl]benzene-1,2-diol
SMILES Oc1cc(O)cc(\C=C\c2ccc(O)c(O)c2)c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H12O4/c15-11-5-10(6-12(16)8-11)2-1-9-3-4-13(17)14(18)7-9/h1-8,15-18H/b2-1+
Formula Weight 244.0736
Chemical Name or Material Piceatannol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.