missing translation for 'onlineSavingsMsg'
Learn More

trans-Permethrinic acid 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30312566
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312566 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312566 Supplier Dr. Ehrenstorfer Supplier No. DREA12507509AL100
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-trans-Permethrinic Acid
CAS 59042-50-1
Analyte or Component Names rac-trans-Permethrinic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H10 Cl2 O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym (1R,3S)-rel-3-(2,2-Dichloroethenyl)-2,2-dimethylcyclopropanecarboxylic Acid, trans-3-(2,2-Dichloroethenyl)-2,2-dimethylcyclopropanecarboxylic Acid, (±)-trans-3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarboxylic Acid, (±)-trans-Cypermethric Acid, (±)-trans-Permethric Acid, trans-(Dichlorovinyl)dimethylcyclopropanecarboxylic Acid, trans-3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropane-1-carboxylic Acid, trans-3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarboxylic Acid, trans-DCCA, trans-Permethrinic Acid, Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, trans-, rel-(1R,3S)-3-(2,2-Dichloroethenyl)-2,2-dimethylcyclopropanecarboxylic acid, (±)-trans-3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarboxylic acid, (±)-trans-Cypermethric acid, (±)-trans-Permethrinic acid, trans-(Dichlorovinyl)dimethylcyclopropanecarboxylic acid, trans-3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropane-1-carboxylic acid, trans-3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarboxylic acid, trans-DCCA, trans-Permethric acid, Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, (1R,3S)-rel-, trans-Permethrinic Acid
InChI Formula InChI=1S/C8H10Cl2O2/c1-8(2)4(3-5(9)10)6(8)7(11)12/h3-4,6H,1-2H3,(H,11,12)/t4-,6+/m1/s1
SMILES CC1(C)[C@H](C=C(Cl)Cl)[C@H]1C(=O)O
IUPAC Name (1R,3S)-3-(2,2-dichlorovinyl)-2,2-dimethyl-cyclopropanecarboxylic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 209.07
Quantity 1 mL
Formula Weight 208.0058 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.