missing translation for 'onlineSavingsMsg'
Learn More

trans-o-Coumaric acid, Dr. Ehrenstorfer

Product Code. 30321707
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321707 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321707 Supplier Dr. Ehrenstorfer Supplier No. DREC11734000
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Coumaric acid
CAS 614-60-8
Analyte or Component Names 2-Coumaric acid
Shipping Condition +20°C
Molecular Formula C9 H8 O3
Certifications/Compliance ISO/IEC 17025
Synonym 2-Propenoic acid, 3-(2-hydroxyphenyl)-, (2E)-, 2-Propenoic acid, 3-(2-hydroxyphenyl)-, (E)-, Cinnamic acid, o-hydroxy-, (E)- (8CI), (2E)-3-(2-Hydroxyphenyl)-2-propenoic acid, (E)-2-Hydroxycinnamic acid, (E)-3-(2-Hydroxyphenyl)acrylic acid, (E)-o-Hydroxycinnamic acid, NSC 32952, o-Hydroxy-trans-cinnamic acid, trans-2-Hydroxycinnamic acid, trans-o-Coumaric acid, trans-o-Hydroxycinnamic acid
InChI Formula InChI=1S/C9H8O3/c10-8-4-2-1-3-7(8)5-6-9(11)12/h1-6,10H,(H,11,12)/b6-5+
Solution Type Neat
SMILES OC(=O)\C=C\c1ccccc1O
IUPAC Name (E)-3-(2-hydroxyphenyl)prop-2-enoic acid
Industry Type Environmental Testing
Molecular Weight (g/mol) 164.16
Quantity 100 mg
Formula Weight 164.0473 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.