missing translation for 'onlineSavingsMsg'
Learn More

trans-beta-Methylstyrene, 97%, stabilized

Product Code. 10301424 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10301424 50 g Glass bottle 50g
10647774 1 g Glass bottle 1g
10446790 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10301424 Supplier Thermo Scientific Acros Supplier No. 150150500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 873-66-5 | C9H10 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-66-5
Molecular Formula C9H10
Molecular Weight (g/mol) 118.18
MDL Number MFCD00009280
InChI Key QROGIFZRVHSFLM-QHHAFSJGSA-N
Synonym beta-methylstyrene, trans-beta-methylstyrene, 1-phenylpropene, isoallylbenzene, propenylbenzene, trans-1-phenyl-1-propene, 1-propenylbenzene, trans-propenylbenzene, 1-phenyl-1-propene, benzene, 1-propenyl
PubChem CID 252325
IUPAC Name [(E)-prop-1-enyl]benzene
SMILES CC=CC1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 0.9110g/mL
Boiling Point 175°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCH3
Refractive Index 1.5480 to 1.55
Quantity 50 g
Beilstein 05, 481
Specific Gravity 0.911
Stabilizer 0.01% BHT
Formula Weight 118.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material trans-β-Methylstyrene, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • trans-ß-Methylstyrene, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.