missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ trans-ISRIB

Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
trans-ISRIB is a potent integrated stress response (ISR) inhibitor; reverses the effects of eIF2α phosphorylation (IC50 = 5 nM) and restores cell translation capacity. Acts downstream of PERK, targeting interactions between eIF2α kinases and elF2B. Blocks ATF4 production without altering the IRE1 or ATF6 responses in vitro. Displays memory enhancement in hippocampus-dependent spatial and fear-associated learning in rodents. trans-ISRIB promotes survival of pluripotent stem cells in culture when used in combination with Chroman 1 (Cat. No. 7163), Emricasan (Cat. No. 7310), and Polyamine Supplement x1000 (lyophilized) (Cat. No. 7739), a combination known as CEPT. When used as part of the CEPT cocktail, trans-ISRIB improves survival of differentiated cells following cryopreservation. Brain penetrant.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | trans-ISRIB |
| Purity | >99% |
| Molecular Weight (g/mol) | 451.34 |
| Inhibitors | trans-ISRIB |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction