missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-Indole-3-acrylic Acid, TRC
Specifications
Specifications
| CAS | 29953-71-7 |
| Molecular Formula | C11H9NO2 |
| Quantity | 250 mg |
| Synonym | (2E)-3-(1H-Indol-3-yl)-2-propenoic Acid, (E)-Indole-3-acrylic Acid, (E)-3-(1H-Indol-3-yl)-2-propenoic Acid, (E)-3-(1H-Indol-3-yl)prop-2-enoic Acid, trans-3-Indoleacrylic Acid, trans-beta-Indoleacrylic Acid; |
| InChI Formula | InChI=1S/C11H9NO2/c13-11(14)6-5-8-7-12-10-4-2-1-3-9(8)10/h1-7,12H,(H,13,14)/b6-5+ |
| SMILES | OC(=O)\C=C\c1c[nH]c2ccccc12 |
| IUPAC Name | (E)-3-(1H-indol-3-yl)prop-2-enoic acid |
| Molecular Weight (g/mol) | 187.19 |
| Formula Weight | 187.0633 |
| Chemical Name or Material | trans-Indole-3-acrylic Acid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?