missing translation for 'onlineSavingsMsg'
Learn More

trans-Feruloylstigmastanol, TRC

Product Code. 30393142
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393142 25 mg
30396545 50 mg
30392816 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393142 Supplier Toronto Research Chemicals Supplier No. TRCF30952025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117857-70-2
Molecular Formula C39H60O4
Molecular Weight (g/mol) 592.89
Synonym Stigmastan-3-ol, 3-(4-hydroxy-3-methoxyphenyl)-2-propenoate, [3beta(E),5alpha]-Sitostanyl Ferulate, Stigmastanol trans-Ferulate, (E)-(3S,5S,8R,9S,10S,13R,14S,17R)-17-((2R,5R)-5-ethyl-6-methylheptan-2-yl)-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-3-yl 3-(4-hydroxy-3-methoxyphenyl)acrylate
SMILES CC[C@H](CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC(=O)\C=C\c5ccc(O)c(OC)c5)C(C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C39H60O4/c1-8-28(25(2)3)12-9-26(4)32-15-16-33-31-14-13-29-24-30(19-21-38(29,5)34(31)20-22-39(32,33)6)43-37(41)18-11-27-10-17-35(40)36(23-27)42-7/h10-11,17-18,23,25-26,28-34,40H,8-9,12-16,19-22,24H2,1-7H3/b18-11+/t26-,28-,29+,30+,31+,32-,33+,34+,38+,39-/m1/s1
Formula Weight 592.4492
Chemical Name or Material trans-Feruloylstigmastanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.