missing translation for 'onlineSavingsMsg'
Learn More

trans-Ferulaldehyde, TRC

Product Code. 30390211
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390211 500 mg -
30392657 100 mg -
30400592 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390211 Supplier Toronto Research Chemicals Supplier No. TRCH973078500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20649-42-7
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.18
Synonym (2E)-3-(4-Hydroxy-3-methoxyphenyl)-2-propenal, (2E)-4-Hydroxy-3-methoxycinnamaldehyde, (E)-3-(4-Hydroxy-3-methoxyphenyl)acrylaldehyde, (E)-3-(4-Hydroxy-m-methoxyphenyl)prop-2-enal, (E)-Coniferaldehyde, (E)-Coniferyl aldehyde, (E)-Ferulaldehyde, 4-Hydroxy-3-methoxy-trans-cinnamaldehyde, E-Coniferyl aldehyde, trans-Coniferaldehyde, trans-Coniferyl Aldehyde
IUPAC Name (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enal
SMILES COc1cc(\C=C\C=O)ccc1O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H10O3/c1-13-10-7-8(3-2-6-11)4-5-9(10)12/h2-7,12H,1H3/b3-2+
Formula Weight 178.063
Chemical Name or Material trans-Ferulaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.