missing translation for 'onlineSavingsMsg'
Learn More

trans-(E)-Flupentixol Dihydrochloride, TRC

Product Code. 30396161
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396161 100 mg
30397342 50 mg
30405878 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396161 Supplier Toronto Research Chemicals Supplier No. TRCF597981100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51529-02-3
Molecular Formula C23H27Cl2F3N2OS
Molecular Weight (g/mol) 507.44
SMILES Cl.Cl.OCCN1CCN(CCC=C2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C23H25F3N2OS.2ClH/c24-23(25,26)17-7-8-22-20(16-17)18(19-4-1-2-6-21(19)30-22)5-3-9-27-10-12-28(13-11-27)14-15-29;;/h1-2,4-8,16,29H,3,9-15H2;2*1H/b18-5+;;
Chemical Name or Material trans-(E)-Flupentixol Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.