missing translation for 'onlineSavingsMsg'
Learn More

trans-Dichlorobis(triphenylphosphine)palladium(II), Premion™, 99.95% (metals basis), Pd 14.7% min, Thermo Scientific Chemicals

Catalog Number 11379617
Click to view available options
Quantity:
1 g
5 g
25 g
13965-03-2
C36H30Cl2P2Pd
701.90
MFCD00009593
YNHIGQDRGKUECZ-UHFFFAOYSA-L
bis triphenylphosphine palladium ii dichloride, bis triphenylphosphine palladium ii chloride, dichlorobis triphenylphosphine palladium, bis triphenylphosphine palladium chloride, bis triphenylphosphine palladiuml ii dichloride, palladium 2+ ; triphenylphosphane; dichloride, trans-dichlorobis triphenylphosphine palladium ii 1g
131664180
ethane;methane;palladium(2+);triphenylphosphane;dichloride
[Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
13965-03-2
C36H30Cl2P2Pd
701.90
MFCD00009593
YNHIGQDRGKUECZ-UHFFFAOYSA-L
bis triphenylphosphine palladium ii dichloride, bis triphenylphosphine palladium ii chloride, dichlorobis triphenylphosphine palladium, bis triphenylphosphine palladium chloride, bis triphenylphosphine palladiuml ii dichloride, palladium 2+ ; triphenylphosphane; dichloride, trans-dichlorobis triphenylphosphine palladium ii 1g
131664180
ethane;methane;palladium(2+);triphenylphosphane;dichloride
[Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

As a catalyst of choice in hydrogenation, hydrosilation, carbonylation, oxidation, elimination, reduction, allyl aceate isomerization, dimerization of 1,3-dienes, and C-C bond formation such as Negishi coupling, Suzuki coupling, Kumada coupling, Sonogashira coupling, Heck coupling and Sonogashira-Hagihara coupling reactions. Used in the preparation of tetrakis(triphenylphosphine)palladium, condensed heterocycles such as furoquinolines, diphenylacetylenes, and several other compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼310°C (decomposition)
Color Yellow
Odor Odorless
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water. Soluble in benzene,and toluene.
Formula Weight 701.91
Percent Purity 99.95%
Grade Premion™
Concentration or Composition (by Analyte or Components) Pd 14.7% min.
Physical Form Powder
Assay (metals basis)
Chemical Name or Material trans-Dichlorobis(triphenylphosphine)palladium(II)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'