missing translation for 'onlineSavingsMsg'
Learn More

trans-Diamminedichloroplatinum(II), Pt 64.5% min

Product Code. 11365978
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365978 5 g 5g
11355978 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365978 Supplier Thermo Scientific Alfa Aesar Supplier No. 010472.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 14913-33-8 | Cl2H6N2Pt | 300.05 g/mol

trans-Diamminedichloroplatinum(II) acts as a reversible RNA-protein cross-linking agent. It is also used in plating baths. It reacts with DNA after hydrolysis to get both intra and interstrand crosslinks. These crosslinks appear to impair replication and transcription of DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14913-33-8
Molecular Formula Cl2H6N2Pt
Molecular Weight (g/mol) 300.05
MDL Number MFCD00011623
InChI Key LXZZYRPGZAFOLE-UHFFFAOYSA-L
Synonym trans-dichlorodiamineplatinum ii, azane; dichloroplatinum, h6cl2n2pt, ia call, ddp-h, platinol tn, cl.2h3n.pt, platinum ii chloride diamine, spectrum1502107, cisplatin jp17/usp/inn
PubChem CID 5702198
ChEBI CHEBI:27899
IUPAC Name azane;dichloroplatinum
SMILES N.N.Cl[Pt]Cl

Specifications

Melting Point 340°C (dec.)
Color Yellow
Physical Form Crystalline
Quantity 5 g
Linear Formula PtCl2(NH3)2
Solubility Information Soluble in dimethylsulfoxide and dimethylformamide. Slightly soluble in water.
Formula Weight 300.06
Concentration or Composition (by Analyte or Components) Pt 64.5% min.
Odor Odorless
Chemical Name or Material trans-Diamminedichloroplatinum(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.