missing translation for 'onlineSavingsMsg'
Learn More

trans-Crotonyl chloride, 90%, tech.

Product Code. 10246010 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10246010 5 g Glass bottle 5g
10256010 100 g Glass bottle 100g
10321344 500 g Glass bottle 500g
10311344 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10246010 Supplier Thermo Scientific Acros Supplier No. 155910050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 625-35-4 | C4H5ClO | 104.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-35-4
Molecular Formula C4H5ClO
Molecular Weight (g/mol) 104.54
MDL Number MFCD00000734
InChI Key RJUIDDKTATZJFE-NSCUHMNNSA-N
Synonym crotonyl chloride, 2-butenoyl chloride, crotonoyl chloride, e-but-2-enoyl chloride, e-2-butenoyl chloride, crotonic acid chloride, 2e-but-2-enoyl chloride, 2-butenoyl chloride, e, 2-butenoyl chloride, 2e, trans-2-butenoyl chloride
PubChem CID 643131
IUPAC Name (E)-but-2-enoyl chloride
SMILES CC=CC(=O)Cl

Specifications

Color Colorless to Brown
Density 1.0900g/mL
Boiling Point 120°C to 123°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula CH3CH=CHCOCl
Refractive Index 1.4585 to 1.4615
Quantity 5 g
Beilstein 02, 411
Fieser 13,208
Specific Gravity 1.09
Solubility Information Solubility in water: decomposes
Formula Weight 104.54
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material trans-Crotonyl chloride, tech., 90%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • trans-Crotonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.