missing translation for 'onlineSavingsMsg'
Learn More

trans-Clomiphene Citrate, TRC

Product Code. 30359193
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359193 10 mg 10mg
30362138 100 mg 100mg
30365526 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359193 Supplier Toronto Research Chemicals Supplier No. TRCC58704510MG
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7599-79-3
Molecular Formula C32H36ClNO8
Molecular Weight (g/mol) 598.08
Synonym 2-[4-[(1E)-2-Chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine 2-Hydroxy-1,2,3-propanetricarboxylate, (E)-Clomiphene Citrate, Clomiphene B Citrate, Enclomid, Enclomiphene Citrate, trans-Clomiphene Citrate
IUPAC Name 2-[4-[(E)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(CC(O)=O)(C(O)=O)CC(O)=O.Cl/C(C1=CC=CC=C1)=C(C2=CC=CC=C2)/C3=CC=C(OCCN(CC)CC)C=C3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C26H28ClNO.C6H8O7/c1-3-28(4-2)19-20-29-24-17-15-22(16-18-24)25(21-11-7-5-8-12-21)26(27)23-13-9-6-10-14-23;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-18H,3-4,19-20H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/b26-25+;
Formula Weight 597.2129
Chemical Name or Material trans-Clomiphene Citrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.