missing translation for 'onlineSavingsMsg'
Learn More

trans-Cinnamoyl chloride, 97%

Product Code. 11419872
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419872 2500 g 2500g
11499862 100 g 100g
11409872 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419872 Supplier Thermo Scientific Alfa Aesar Supplier No. A12016.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17082-09-6 | C9H7ClO | 166.604 g/mol

The derivative of this compound, trans-Cinnamoylimidazole is involved in the synthesis of trans-Cinnamoyl-Papain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The derivative of this compound, trans-Cinnamoylimidazole is involved in the synthesis of trans-Cinnamoyl-Papain.

Solubility
Reacts with water.

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases. Moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17082-09-6
Molecular Formula C9H7ClO
Molecular Weight (g/mol) 166.60
MDL Number MFCD00000732
InChI Key WOGITNXCNOTRLK-VOTSOKGWSA-N
Synonym cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride
PubChem CID 5354261
IUPAC Name (2E)-3-phenylprop-2-enoyl chloride
SMILES ClC(=O)\C=C\C1=CC=CC=C1

Specifications

Melting Point 35°C to 37°C
Density 1.162
Boiling Point 256°C to 258°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.5845
Quantity 2500 g
UN Number UN3261
Beilstein 606265
Sensitivity Moisture sensitive
Merck Index 14,2301
Solubility Information Reacts with water.
Formula Weight 166.61
Percent Purity 97%
Chemical Name or Material trans-Cinnamoyl chloride
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.