missing translation for 'onlineSavingsMsg'
Learn More

trans-Cinnamaldehyde, TRC

Product Code. 30008900
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008900 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30008900 Supplier Toronto Research Chemicals Supplier No. TRCC4420205G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14371-10-9
Molecular Formula C9 H8 O
Molecular Weight (g/mol) 132.16
Synonym (E)-Cinnamaldehyde, (2E)-3-phenyl-2-propenal
IUPAC Name (E)-3-phenylprop-2-enal
SMILES O=C\C=C\c1ccccc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H8O/c10-8-4-7-9-5-2-1-3-6-9/h1-8H/b7-4+
Formula Weight 132.0575
Chemical Name or Material trans-Cinnamaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.