missing translation for 'onlineSavingsMsg'
Learn More

trans-beta-Nitrostyrene, 97+%

Product Code. 11393496
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393496 5 g 5g
11303506 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393496 Supplier Thermo Scientific Alfa Aesar Supplier No. L14367.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5153-67-3 | C8H7NO2 | 149.15 g/mol

trans-beta-Nitrostyrene is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-beta-Nitrostyrene is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5153-67-3
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00007402
InChI Key PIAOLBVUVDXHHL-VOTSOKGWSA-N
Synonym trans-beta-nitrostyrene, 2-nitrovinyl benzene, e-2-nitrovinyl benzene, beta-nitrostyrene, 2-nitro-1-phenylethylene, e-beta-nitrostyrene, 2-nitroethenyl benzene, 1-phenyl-2-nitroethene, beta-nitrosytrene, benzene, 2-nitroethenyl
PubChem CID 5284459
IUPAC Name [(E)-2-nitroethenyl]benzene
SMILES [O-][N+](=O)\C=C\C1=CC=CC=C1

Specifications

Melting Point 55°C to 58°C
Boiling Point 250°C to 260°C
Quantity 5 g
Beilstein 1210066
Solubility Information Insoluble in water.
Formula Weight 149.15
Percent Purity 98%
Chemical Name or Material trans-beta-Nitrostyrene
compliance-icons
Product Identifier
  • trans-beta-Nitrostyrene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: trans-.beta.-Nitrostyrene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.