missing translation for 'onlineSavingsMsg'
Learn More

Trans-Anethole, MedChemExpress

Product Code. 16429066
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16429066 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16429066 Supplier Medchem Express Supplier No. HYN0367
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Trans-Anethole ((E)-Anethole), a phenylpropene derivative isolated from Pimpinella, shows estrogenic activity at lower concentrations and cytotoxic at higher concentrations in cancer cell lines. Trans-Anethole ((E)-Anethole) contributes a large component of the odor and flavor of anise and fennel, anise myrtle, liquorice, camphor, magnolia blossoms, and star anise.

Trans-Anethole ((E)-Anethole), a phenylpropene derivative isolated from Pimpinella, shows estrogenic activity at lower concentrations and cytotoxic at higher concentrations in cancer cell lines. Trans-Anethole ((E)-Anethole) contributes a large component of the odor and flavor of anise and fennel, anise myrtle, liquorice, camphor, magnolia blossoms, and star anise.

Specifications

Chemical Name or Material Trans-Anethole
CAS 4180-23-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C10H12O
Quantity 100 mg
Synonym (E)​-Anethole
Solubility Information DMSO : 100 mg/mL (674.76 mM; Need ultrasonic) ∣H2O : 1 mg/mL (6.75 mM; ultrasonic and warming and heat to 80°C)
SMILES COC1=CC=C(/C=C/C)C=C1
Molecular Weight (g/mol) 148.2
Formula Weight 148.2
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Informativa sulla privacy.