missing translation for 'onlineSavingsMsg'
Learn More

trans-Aconitic Acid, TRC

Product Code. 30138158
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138158 5 g 5g
30136451 1 g 1g
30003763 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138158 Supplier Toronto Research Chemicals Supplier No. TRCA1898855G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4023-65-8
Molecular Formula C6 H6 O6
Molecular Weight (g/mol) 174.11
Synonym 1-Propene-1,2,3-tricarboxylic acid, (1E)-, 1-Propene-1,2,3-tricarboxylic acid, (E)- (8CI), Aconitic acid, trans- (6CI), (1E)-1-Propene-1,2,3-tricarboxylic acid, (1E)-Prop-1-ene-1,2,3-tricarboxylic acid, (E)-1-Propene-1,2,3-tricarboxylic acid, (E)-Aconitic acid, trans-1-Propene-1,2,3-tricarboxylic acid, trans-Aconitic acid
IUPAC Name (E)-prop-1-ene-1,2,3-tricarboxylic acid
SMILES OC(=O)C\C(=C/C(=O)O)\C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H6O6/c7-4(8)1-3(6(11)12)2-5(9)10/h1H,2H2,(H,7,8)(H,9,10)(H,11,12)/b3-1+
Formula Weight 174.0164
Chemical Name or Material trans-Aconitic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.