Learn More
trans-Aconitic acid, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
It plays an important role as an antimicrobial agent that blocks the transformation of Leishmania donovani amastigotes to prostigotes. trans-Aconitic acid is also used in synthesis of aconitate esters. It may be used for cross-linking polyvinyl alcohol film for preparing composite nanofiltration membranes.
Solubility
Soluble in water at 20°C 400g/L. Slightly soluble in ether.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | ∼190°C (decomposition) |
| Odor | Odorless |
| Quantity | 100 g |
| Merck Index | 14,117 |
| Solubility Information | Soluble in water at 20°C 400g/L. Slightly soluble in ether. |
| Formula Weight | 174.11 |
| Percent Purity | 98% |
| Chemical Name or Material | trans-Aconitic acid |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.