missing translation for 'onlineSavingsMsg'
Learn More

trans-5-Decene, 97%

Product Code. 11372936
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372936 5 g 5g
11362936 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372936 Supplier Thermo Scientific Alfa Aesar Supplier No. L10689.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7433-56-9 | C10H20 | 140.27 g/mol

trans-5-Decene has been used to characterize the effect of cyclization and size on the stability of Criegee intermediates formed in ozonolysis. It has been added to phenoxathiin cation radical in acetonitrile solution to form bis(10-phenoxathiiniumyl) alkane adducts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-5-Decene has been used to characterize the effect of cyclization and size on the stability of Criegee intermediates formed in ozonolysis. It has been added to phenoxathiin cation radical in acetonitrile solution to form bis(10-phenoxathiiniumyl) alkane adducts.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7433-56-9
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00009508
InChI Key UURSXESKOOOTOV-MDZDMXLPSA-N
Synonym trans-5-decene, e-5-decene, 5-decene, e, trans-dec-5-ene, 5e-decene, 5-decene, cis-5-decene, 5-decene, 5e, 5-trans-decene, 5e dec-5-ene
PubChem CID 637820
IUPAC Name (E)-dec-5-ene
SMILES CCCC\C=C\CCCC

Specifications

Density 0.74
Boiling Point 171°C to 172°C
Flash Point 46°C (114°F)
Refractive Index 1.424
Quantity 5 g
UN Number UN3295
Beilstein 1719487
Solubility Information Insoluble in water.
Formula Weight 140.27
Percent Purity 97%
Chemical Name or Material trans-5-Decene
Show More Show Less
compliance-icons
Product Identifier
  • trans-5-Decene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: trans-Dec-5-ene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.