missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Octene, 97%

Product Code. 11344838
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344838 10 g 10g
11354838 2 g 2g
11334838 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11344838 Supplier Thermo Scientific Alfa Aesar Supplier No. 043155.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14850-23-8 | C8H16 | 112.216 g/mol

It is used in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14850-23-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00009476
InChI Key IRUCBBFNLDIMIK-BQYQJAHWSA-N
Synonym trans-4-octene, e-4-octene, trans-n-4-octene, e-oct-4-ene, 4-octene, e, 4-octene, 4-octene, 4e, unii-fkz5737b1w, n-trans-4-octene, 4e-4-octene
PubChem CID 5357253
IUPAC Name (E)-oct-4-ene
SMILES CCCC=CCCC

Specifications

Density 0.715 g/mL
Boiling Point 122°C to 123°C
Flash Point 21°C (70°F)
Odor Characteristic
Refractive Index 1.412
Quantity 10 g
UN Number UN3295
Beilstein 1719104
Solubility Information Sparingly soluble in water(10.7,mg/L).
Formula Weight 112.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material trans-4-Octene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.