missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Methyl-beta-nitrostyrene, 98%, Thermo Scientific Chemicals

Product Code. 11477746
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

trans-4-Methyl-β-nitrostyrene may be used as a reagent in the synthesis of N-benzylpyrrolomorphinans. It is also used as an organic intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Methyl-β-nitrostyrene may be used as a reagent in the synthesis of N-benzylpyrrolomorphinans. It is also used as an organic intermediate for pharmaceutical.

Solubility
Sparingly Soluble in water (0.070 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5153-68-4
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00024828
InChI Key JSPNBERPFLONRX-VOTSOKGWSA-N
Synonym 1-methyl-4-2-nitrovinyl benzene, trans-4-methyl-beta-nitrostyrene, e-1-methyl-4-2-nitrovinyl benzene, 4-methyl-b-nitrostyrene, 4-methyl-beta-nitrostyrene, 1-methyl-4-2-nitroethenyl benzene, 1-methyl-4-e-2-nitroethenyl benzene, 4-methyl-.beta.-nitrostyrene, 4-methyl-.omega.-nitrostyrene, e-beta-nitro-4-methylstyrene
PubChem CID 674135
IUPAC Name 1-methyl-4-[(E)-2-nitroethenyl]benzene
SMILES CC1=CC=C(C=C1)C=C[N+](=O)[O-]

Specifications

Melting Point 100°C to 104°C
Quantity 5 g
Beilstein 1864209
Solubility Information Sparingly Soluble in water (0.070g/L) (25°C).
Formula Weight 163.18
Percent Purity 98%
Chemical Name or Material trans-4-Methyl-beta-nitrostyrene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'