missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Methyl-1-cyclohexanecarboxylic acid, 98+%, Thermo Scientific™

Product Code. 10506494 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10506494 5 g Glass bottle 5g
10274774 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10506494 Supplier Thermo Scientific Acros Supplier No. 427500050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13064-83-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00074943,MFCD00074909,MFCD20617670
InChI Key QTDXSEZXAPHVBI-UHFFFAOYSA-N
Synonym trans-4-methylcyclohexanecarboxylic acid, 4-methylcyclohexanecarboxylic acid, 4-methyl-1-cyclohexanecarboxylic acid, cis-4-methylcyclohexanecarboxylic acid, unii-70f0i2amtv, unii-m7dsk28350, trans-4-methyl-1-cyclohexanecarboxylic acid, trans-4-methyl hexahydrobenzoic acid, trans-4-methylcyclohexane carboxylic acid, 70f0i2amtv
PubChem CID 20330
IUPAC Name 4-methylcyclohexane-1-carboxylic acid
SMILES CC1CCC(CC1)C(O)=O

Specifications

Melting Point 109.0°C to 111.0°C
Boiling Point 245.0°C
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 5 g
Formula Weight 142.2
Percent Purity 98+%
Chemical Name or Material trans-4-Methyl-1-cyclohexanecarboxylic acid
compliance-icons
Product Identifier
  • trans-4-Methyl-1-cyclohexanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.