missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Methoxycinnamaldehyde, 98+%

Product Code. 11465233
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465233 10 g 10g
11475233 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465233 Supplier Thermo Scientific Alfa Aesar Supplier No. A12839.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 24680-50-0 | C10H10O2 | 162.19 g/mol

trans-4-Methoxycinnamaldehyde is used as an intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Methoxycinnamaldehyde is used as an intermediate in chemical research.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24680-50-0
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00017343
InChI Key AXCXHFKZHDEKTP-NSCUHMNNSA-N
Synonym 4-methoxycinnamaldehyde, 3-4-methoxyphenyl acrylaldehyde, p-methoxycinnamaldehyde, e-3-4-methoxyphenyl acrylaldehyde, 3-4-methoxyphenyl-2-propenal, trans-4-methoxycinnamaldehyde, p-methoxy cinnamaldehyde, p-methoxycinnamic aldehyde, trans-p-methoxycinnamaldehyde, cinnamaldehyde, p-methoxy
PubChem CID 641294
ChEBI CHEBI:27952
IUPAC Name (E)-3-(4-methoxyphenyl)prop-2-enal
SMILES COC1=CC=C(\C=C\C=O)C=C1

Specifications

Melting Point 56°C to 60°C
Boiling Point 160°C (3 mmHg)
Flash Point >110°C (230°F)
Odor Pleasant
Quantity 10 g
Beilstein 1447540
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 162.19
Percent Purity ≥98%
Chemical Name or Material trans-4-Methoxycinnamaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.