missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxystilbene, 98%

Product Code. 10224880 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10224880 5 g 5g
10284920 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10224880 Supplier Thermo Scientific Acros Supplier No. 121930050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6554-98-9 | C14H12O | 196.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6554-98-9
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00002386
InChI Key QVLMUEOXQBUPAH-VOTSOKGWSA-N
Synonym trans-4-hydroxystilbene, 4-hydroxystilbene, 4-styrylphenol, 4-stilbenol, e-4-stilbenol, 4-2-phenylethenyl phenol, 4-2-phenylvinyl phenol, 4-e-2-phenylethenyl phenol, e-4-hydroxystilbene, stilben-4-ol
PubChem CID 5284650
ChEBI CHEBI:35101
IUPAC Name 4-[(E)-2-phenylethenyl]phenol
SMILES C1=CC=C(C=C1)C=CC2=CC=C(C=C2)O

Specifications

Melting Point 185°C to 189°C
Assay Percent Range 96.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula C6H5CH=CHC6H4OH
Quantity 5 g
Beilstein 06,693
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,chloroform,benzene,,soluble in acetic acid
Formula Weight 196.25
Percent Purity 98%
Chemical Name or Material trans-4-Hydroxystilbene, 98%
compliance-icons
Product Identifier
  • trans-4-Hydroxystilbene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.