missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxycyclohexylcarboxylic Acid, TRC

Product Code. 30397869
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397869 1 g 1g
30399778 500 mg -
30391962 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397869 Supplier Toronto Research Chemicals Supplier No. TRCH8269001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3685-26-5
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
Synonym 4-Hydroxycylohexanecarboxylic Acid, (1r,2r)-4-Hydroxycyclohexan-1-carboxylic Acid
IUPAC Name 4-hydroxycyclohexane-1-carboxylic acid
SMILES O[C@@H]1CC[C@H](CC1)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H12O3/c8-6-3-1-5(2-4-6)7(9)10/h5-6,8H,1-4H2,(H,9,10)
Formula Weight 144.0786
Chemical Name or Material trans-4-Hydroxycyclohexylcarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.