missing translation for 'onlineSavingsMsg'
Learn More

trans 4-Hydroxy-L-proline beta-Naphthylamide, TRC

Product Code. 30407058
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407058 500 mg 500mg
30398784 250 mg -
30397579 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407058 Supplier Toronto Research Chemicals Supplier No. TRCH952390500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3326-64-5
Molecular Formula C15 H16 N2 O2
Molecular Weight (g/mol) 256.3
Synonym Trans 4-Hydroxy-L-Proline -B-Naphthylamide
IUPAC Name (2S,4R)-4-hydroxy-N-naphthalen-2-ylpyrrolidine-2-carboxamide
SMILES O[C@H]1CN[C@@H](C1)C(=O)Nc2ccc3ccccc3c2

Specifications

InChI Formula InChI=1S/C15H16N2O2/c18-13-8-14(16-9-13)15(19)17-12-6-5-10-3-1-2-4-11(10)7-12/h1-7,13-14,16,18H,8-9H2,(H,17,19)/t13-,14+/m1/s1
Quantity 500 mg
Formula Weight 256.1212
Shipping Condition Room Temperature
Chemical Name or Material trans 4-Hydroxy-L-proline beta-Naphthylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.