missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals trans-4-Hydroxy-L-proline, 99+%

Product Code. 10336470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10336470 100 g Glass bottle 100g
10326470 10 g Glass bottle 10g
10607662 25 g Glass bottle 25g
11926601 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10336470 Supplier Thermo Scientific Chemicals Supplier No. 121781000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51-35-4 | C5H9NO3 | 131.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-35-4
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
InChI Key PMMYEEVYMWASQN-DMTCNVIQSA-N
Synonym trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro
PubChem CID 5810
ChEBI CHEBI:18095
IUPAC Name (2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES C1C(CNC1C(=O)O)O

Specifications

Melting Point 274.0°C to 275.0°C
Color White
Loss on Drying 0.2% (105°C, 3 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 100 g
Beilstein 22,191
Merck Index 15,4878
Specific Rotation Condition - 75.50 (20.00°C c=5,H2O)
Specific Rotation − 75.50
Solubility Information Solubility in water: 357.8g/L (20° c). Other solubilities: slightly soluble in alcohol, insoluble in ether
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Formula Weight 131.13
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material trans-4-Hydroxy-L-proline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.