missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxy-1-L-phenylalanyl-L-proline, TRC

Product Code. 30403593
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403593 50 mg -
30396820 10 mg -
30390635 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403593 Supplier Toronto Research Chemicals Supplier No. TRCH24555050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90965-82-5
Molecular Formula C14 H18 N2 O4
Molecular Weight (g/mol) 278.304
Synonym (4R)-L-Phenylalanyl-4-hydroxy-L-proline
IUPAC Name (2S,4R)-1-[(2S)-2-amino-3-phenylpropanoyl]-4-hydroxypyrrolidine-2-carboxylic acid
SMILES N[C@@H](Cc1ccccc1)C(=O)N2C[C@H](O)C[C@H]2C(=O)O

Specifications

InChI Formula InChI=1S/C14H18N2O4/c15-11(6-9-4-2-1-3-5-9)13(18)16-8-10(17)7-12(16)14(19)20/h1-5,10-12,17H,6-8,15H2,(H,19,20)/t10-,11+,12+/m1/s1
Quantity 50 mg
Formula Weight 278.127
Shipping Condition Room Temperature
Chemical Name or Material trans-4-Hydroxy-1-L-phenylalanyl-L-proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.