missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Decenal, 96%, may cont. ca 7% cis-isomer

Product Code. 11367565
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11367565

Brand: Thermo Scientific Alfa Aesar L05715.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65405-70-1 | C10H18O | 154.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65405-70-1
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00065447
InChI Key CWRKZMLUDFBPAO-VOTSOKGWSA-N
Synonym trans-4-decen-1-al, trans-4-decenal, e-4-decenal, e-dec-4-enal, 4-decenal, e, 4-decenal, unii-kz74b5801j, 4-decene-1-al, dec-4-enal, fema no. 3264
PubChem CID 5702654
IUPAC Name (E)-dec-4-enal
SMILES CCCCCC=CCCC=O

Specifications

Density 0.842
Boiling Point 80°C (4 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.445
Quantity 5 g
Beilstein 4230058
Formula Weight 154.25
Percent Purity 96%
Assay may cont. ≈7% cis-isomer
Chemical Name or Material trans-4-Decenal

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.