missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Bromo-beta-nitrostyrene, 99%

Product Code. p-4323862 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11904591

Brand: Thermo Scientific Acros 440160010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5153-71-9 | C8H6BrNO2 | 228.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5153-71-9
Molecular Formula C8H6BrNO2
Molecular Weight (g/mol) 228.05
InChI Key LSGVHLGCJIBLMB-AATRIKPKSA-N
Synonym 1-bromo-4-2-nitrovinyl benzene, e-1-bromo-4-2-nitrovinyl benzene, 1-bromo-4-e-2-nitroethenyl benzene, trans-4-bromo-beta-nitrostyrene, benzene, 1-bromo-4-1e-2-nitroethenyl, beta-nitro-4-bromostyrene, 4-bromo-.beta.-nitrostyrene, e-4-bromo-beta-nitrostyrene, 1-bromo-4-e-2-nitrovinyl benzene
PubChem CID 688204
IUPAC Name 1-bromo-4-[(E)-2-nitroethenyl]benzene
SMILES C1=CC(=CC=C1C=C[N+](=O)[O-])Br

Specifications

Melting Point 148.0°C to 152.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 228.05
Percent Purity 99%
Physical Form Crystalline Powder or Powder
Chemical Name or Material trans-4-Bromo-β-nitrostyrene
compliance-icons
Product Identifier
  • trans-4-Bromo-beta-nitrostyrene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.