missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Boc-aminomethyl)cyclohexanemethanol, 97%

Product Code. 15479268
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15479268

Brand: Thermo Scientific Alfa Aesar H62798.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 172348-63-9 | C13H25NO3 | 243.347 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172348-63-9
Molecular Formula C13H25NO3
Molecular Weight (g/mol) 243.347
MDL Number MFCD06657666
InChI Key KSXPGASLEFGROT-UHFFFAOYSA-N
Synonym tert-butyl trans-4-hydroxymethylcyclohexylmethyl-carbamate, trans-4-boc-aminomethyl-1-hydroxymethyl-cyclohexane, tert-butyl trans-4-hydroxymethyl cyclohexyl methyl carbamate, tert-butyl trans-4-hydroxymethylcyclohexylmethyl carbamate, tert-butyl 4-hydroxymethyl cyclohexyl methylcarbamate, ksxpgaslefgrot-xypyzodxsa-n, tert-butyl 4-hydroxymethyl cyclohexyl methyl carbamate, trans-4-n-tert-butoxycarbonylaminomethyl cyclohexylmethanol, n-tert-butoxycarbonyl-trans-4-aminomethyl cyclohexanemethanol
PubChem CID 18397396
IUPAC Name tert-butyl N-[[4-(hydroxymethyl)cyclohexyl]methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1CCC(CC1)CO

Specifications

Quantity 5 g
Formula Weight 243.34
Percent Purity 97%
Chemical Name or Material trans-4-(Boc-aminomethyl)cyclohexanemethanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.