missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Boc-amino)cyclohexylamine, 97%

Product Code. 15419218
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419218 100 g 100g
15499208 5 g 5g
15409218 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15419218 Supplier Thermo Scientific Alfa Aesar Supplier No. H62734.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 177906-48-8 | C11H22N2O2 | 214.31 g/mol

trans-4-(Boc-amino)cyclohexylamine is used in the preparation of orally bioavailable and selective V1A receptor antagonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexylamine is used in the preparation of orally bioavailable and selective V1A receptor antagonist.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177906-48-8
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.31
MDL Number MFCD01076211,MFCD03001719,MFCD03844604
InChI Key FEYLUKDSKVSMSZ-UHFFFAOYSA-N
Synonym trans-n-boc-1,4-cyclohexanediamine, n-boc-trans-1,4-cyclohexanediamine, 1-n-boc-cis-1,4-cyclohexyldiamine, tert-butyl 4-aminocyclohexyl carbamate, n-boc-1,4-cyclohexanediamine, tert-butyl n-4-aminocyclohexyl carbamate, tert-butyl cis-4-aminocyclohexyl carbamate, 1-boc-amino-1,4-cyclohexanediamine, tert-butyl 4-aminocyclohexylcarbamate, cis-n-boc-1,4-cyclohexanediamine
PubChem CID 2756050
IUPAC Name tert-butyl N-(4-aminocyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(N)CC1

Specifications

Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 214.3
Percent Purity 97%
Chemical Name or Material trans-4-(Boc-amino)cyclohexylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.