missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine, 97%, Thermo Scientific Chemicals

Product Code. 15416956
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15416956 250 mg 250mg
15496946 1 g 1g
15406956 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15416956 Supplier Thermo Scientific Alfa Aesar Supplier No. H62034.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine used as intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine used as intermediate for chemical research.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177582-74-0
Molecular Formula C15H22N2O2
Molecular Weight (g/mol) 262.35
MDL Number MFCD06657669
InChI Key AJYOPGOCTHCBTL-UHFFFAOYSA-N
Synonym benzyl trans-4-aminomethyl cyclohexyl carbamate, benzyl trans-4-aminomethylcyclohexylcarbamate, benzyl 4-aminomethyl cyclohexyl carbamate, benzyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, ajyopgocthcbtl-mqmhxkeqsa-n, ajyopgocthcbtl-xbxgtlagsa-n, benzyl 4-aminomethyl cyclohexylcarbamate, benzyl n-4-aminomethyl cyclohexyl carbamate, benzyl cis-4-aminomethyl cyclohexyl carbamate, benzyl trans-4-aminomethylcyclohexyl carbamate
PubChem CID 22253963
IUPAC Name benzyl N-[4-(aminomethyl)cyclohexyl]carbamate
SMILES NCC1CCC(CC1)NC(=O)OCC1=CC=CC=C1

Spécification

Beilstein 7710506
Quantity 250 mg
Formula Weight 262.34
Percent Purity 97%
Chemical Name or Material trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.