missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine, 97%, Thermo Scientific Chemicals

Product Code. 15406956
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15406956 5 g 5g
15416956 250 mg 250mg
15496946 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15406956 Supplier Thermo Scientific Alfa Aesar Supplier No. H62034.06
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine used as intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine used as intermediate for chemical research.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177582-74-0
Molecular Formula C15H22N2O2
Molecular Weight (g/mol) 262.35
MDL Number MFCD06657669
InChI Key AJYOPGOCTHCBTL-UHFFFAOYSA-N
Synonym benzyl trans-4-aminomethyl cyclohexyl carbamate, benzyl trans-4-aminomethylcyclohexylcarbamate, benzyl 4-aminomethyl cyclohexyl carbamate, benzyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, ajyopgocthcbtl-mqmhxkeqsa-n, ajyopgocthcbtl-xbxgtlagsa-n, benzyl 4-aminomethyl cyclohexylcarbamate, benzyl n-4-aminomethyl cyclohexyl carbamate, benzyl cis-4-aminomethyl cyclohexyl carbamate, benzyl trans-4-aminomethylcyclohexyl carbamate
PubChem CID 22253963
IUPAC Name benzyl N-[4-(aminomethyl)cyclohexyl]carbamate
SMILES NCC1CCC(CC1)NC(=O)OCC1=CC=CC=C1

Specifications

Beilstein 7710506
Quantity 5 g
Formula Weight 262.34
Percent Purity 97%
Chemical Name or Material trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.