missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Aminocyclohexanol hydrochloride, 97%

Product Code. 11426057
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11426057

Brand: Thermo Scientific Alfa Aesar B25070.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50910-54-8 | C6H14ClNO | 151.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50910-54-8
Molecular Formula C6H14ClNO
Molecular Weight (g/mol) 151.63
MDL Number MFCD00012566,MFCD07366531,MFCD06410647
InChI Key RKTQEVMZBCBOSB-UHFFFAOYSA-N
Synonym trans-4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hydrochloride, 4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hcl, 4-aminocyclohexan-1-ol hydrochloride, cis-4-amino-cyclohexanol hydrochloride, 4-amino-cyclohexanol hydrochloride, 1r,4r-4-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 4-amino-, hydrochloride, trans-4-aminocyclohexanol hcl
PubChem CID 522619
IUPAC Name 4-aminocyclohexan-1-ol;hydrochloride
SMILES Cl.NC1CCC(O)CC1

Specifications

Melting Point 225°C to 230°C
Quantity 5 g
Beilstein 3909294
Sensitivity Hygroscopic
Formula Weight 151.64
Percent Purity 97%
Chemical Name or Material trans-4-Aminocyclohexanol hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.