missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Aminocyclohexanol, 98+%

Product Code. 11496096
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496096 5 g 5g
11406106 25 g 25g
11416106 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496096 Supplier Thermo Scientific Alfa Aesar Supplier No. B22365.06
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 27489-62-9 | C6H13NO | 115.176 g/mol

trans-4-Aminocyclohexanol is used as raw material in organic synthesis and is an important intermediate in the synthesis of drugs such as Ambroxol hydrochloride. It can react with butyric acid-(2-chloro-ethyl ester) to get butyric acid 4-amino-cyclohexyl ester. This reaction needs catalytic agent Aspergillus niger lipase and solvent 2-methyl-butan-2-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Aminocyclohexanol is used as raw material in organic synthesis and is an important intermediate in the synthesis of drugs such as Ambroxol hydrochloride. It can react with butyric acid-(2-chloro-ethyl ester) to get butyric acid 4-amino-cyclohexyl ester. This reaction needs catalytic agent Aspergillus niger lipase and solvent 2-methyl-butan-2-ol.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27489-62-9
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00067698
InChI Key IMLXLGZJLAOKJN-UHFFFAOYSA-N
Synonym trans-4-aminocyclohexanol, 4-aminocyclohexanol, cis-4-aminocyclohexanol, cis-4-amino-cyclohexanol, cyclohexanol, 4-amino-, trans, 1r,4r-4-aminocyclohexan-1-ol, cyclohexanol, 4-amino, trans-4-hydroxycyclohexylamine, 1r,4r-4-aminocyclohexanol, aminocyclohexanol trans-4
PubChem CID 81293
IUPAC Name 4-aminocyclohexan-1-ol
SMILES C1CC(CCC1N)O

Specifications

Melting Point 110°C to 113°C
Quantity 5 g
UN Number UN3259
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 115.18
Percent Purity ≥98%
Chemical Name or Material trans-4-Aminocyclohexanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.